missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFXN5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | SFXN5 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605085
|
Novus Biologicals
NBP2-99580-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672236
|
Novus Biologicals
NBP2-99580-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SFXN5 Polyclonal antibody specifically detects SFXN5 in Mouse samples. It is validated for Western BlotSpecifications
| SFXN5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| BBG-TCC, sideroflexin 5 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SFXN5 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 94097 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title