Learn More
Tocris Bioscience™ SGC 6870
Description
SGC 6870 is a potent and selective protein arginine methyltransferase 6 (PRMT6) allosteric inhibitor (IC50 = 77 nM). Exhibits selectivity over all other PRMTs and 23 methyltransferases. In HEK293T cells overexpressing PRMT6, inhibits asymmetric dimethylation of H3R2 (IC50 = 0.8 μM) Negative control SGC 6870N (Cat. No. 7184) also available.
Specifications
Specifications
| Chemical Name or Material | (R)-4-(5-Bromothiophene-2-carbonyl)-5-(3,5-dimethylphenyl)-7-methyl-1,3,4,5-tetrahydro-2H-benzo[e][1,4]diazepin-2-one |
| CAS | 2561471-27-8 |
| Quantity | 25 mg |
| Target | Protein Arginine Methyltransferase Inhibitors |
| Molecular Formula | C23H21BrN2O2S |
| Purity | 0.98 |