missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGCD Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142693
This item is not returnable.
View return policy
Description
Additional Information: This antibody is expected to recognize isoform 1 (NP_000328.2) and isoform 3 (NP_001121681.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
Specifications
| SGCD | |
| Polyclonal | |
| Unconjugated | |
| SGCD | |
| 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kDa; CMD1L; CMD1LSG-delta; DAGD; delta-sarcoglycan; Delta-SG; dystrophin associated glycoprotein, delta sarcoglycan; LGMD2F; MGC22567; placental delta sarcoglycan; sarcoglycan delta; sarcoglycan, delta (35kD dystrophin-associated glycoprotein); sarcoglycan, delta (35kDa dystrophin-associated glycoprotein); sarcoglycan, delta (dystrophin-associated glycoprotein); sarcoglycandelta; sarcoglycan-delta; SGCD; SGCDP; SGD; SGdelta; SG-delta | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 497892, 6444 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q92629 | |
| SGCD | |
| Peptide with sequence C-RLPHGSYTPTGTR (aa 244-256). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction