Learn More
Trajan™ Beckman/Altex, Rheodyne, SSI, Valco Valve Replacement Needle
Click to view available options
Volume (Metric):
10 μL
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications
Specifications
| Material | PTFE (Plunger) |
| Description | Gas tight |
| Needle Gauge | 22 G |
| Diameter (Metric) Inner Needle | 0.17 mm |
| Diameter (Metric) Outer Needle | 0.028 mm |
| Volume (Metric) | 10 μL |
| Needle Point Style | LC |