Learn More
SGE™ Beckman/Altex, Rheodyne, SSI, Valco Valves with Fixed Needle
Click to view available options
Description:
HPLC Valve Syringe, Fixed LC Needle
Removable Needle Syringe, Removable Plunger
SuperFlex Plunger, Fixed LC Needle
Unit Size:
Each
Pack of 6
Description
- Fixed needle
- For use with For Rheodyne™ and Valco™ Valves
Specifications
Specifications
| Type | Syringe |
| Needle Gauge | 22 G |
| For Use With (Application) | Beckman/Altex, Rheodyne™, SSI instruments, and valco valves |
| Description | SuperFlex Plunger, Fixed LC Needle |
| Diameter (English) Inner | 0.028 in. |
| Diameter (Metric) Inner | 0.71 mm |
| Volume (Metric) | 10 μL |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur