Learn More
SGE™ BP20 (WAX) Capillary Columns
Click to view available options
Length (Metric):
30 m
60 m
Diameter (Metric) Inner:
0.25 mm
0.32 mm
0.53 mm
Film Thickness:
0.25 μm
0.5 μm
1 μm
Unit Size:
Each
Description
- Very polar polyethylene glycol (PEG) phase
- Cross-linked for stability and washing
- Also ideal for xylene and aromatic isomer
- 0.1 to 1μm film thickness: 20° to 260/280°C
Specifications
Specifications
| Diameter (Metric) Inner | 0.25 mm |
| Stationary Phase | SGE™ BP20 (Wax) |
| For Use With (Application) | Gas Chromatography (GC), Ketones, Aldehydes Analysis, Fragrance Analysis |
| Particle Size | 1 μm |
| Film Thickness | 1 μm |
| Length (Metric) | 30 m |
| Polarity | Polar |