Learn More
Trajan™ Phenyl Deactivated Fused Silica Capillary Tubing
Click to view available options
Diameter (Metric) Inner:
0.22 mm
0.32 mm
0.53 mm
Diameter (Metric) Outer:
0.36 mm
0.43 mm
0.68 mm
Unit Size:
5m
Each
Description
- Chemically inert and thermally stable to 380°C
- Suitable for organic and aqueous solvents
- Ideal for biotechnology applications
Specifications
Specifications
| Type | Capillary Tubing |
| Diameter (Metric) Outer | 0.36 mm |
| Diameter (Metric) Inner | 0.22 mm |
| For Use With (Application) | Organic and Aqueous Solvents |
| Deactivation | Phenyl |
| Overall Length (Metric) | 5 m |