Learn More
Trajan™ Diamond GC Needles for 10 to 50μL Manual Syringes
missing translation for 'orderingAttributeHoverText'
Needle Gauge:
25 G
26 G
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
missing translation for 'unitSize'
Each
Descripción
Improve the sensitivity of your MS analysis:
- Lowest syringe carryover
- Reduced non-specific sample interaction and increased analyte recovery
- Ultra smooth removable needles with choice of surface treatment.
Especificaciones
Especificaciones
| Needle Gauge | 25 G |
| Diameter (English) Inner | 0.0078 in. |
| Diameter (English) Outer | 0.196 in. |
| Diameter (Metric) Inner | 0.2 mm |
| Diameter (Metric) Outer | 0.5 mm |
| Includes | Ultra Smooth Coating |
| Length (English) Needle | 2 in. |
| Product Type | GC Needle |
| Needle Point Style | Bevel |
| For Use With (Equipment) | Manual Syringe (10 to 50μL) |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido