Learn More
Trajan™ Diamond GC Needles for 10 to 50μL Manual Syringes
Product Code. 13157916
Click to view available options
Needle Gauge:
25 G
26 G
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Unit Size:
Each
Product Code. 13157916
Brand: Trajan™ 0355310
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond GC Needles for 10 to 50μL Manual Syringes are MS ready and designed to improve the sensitivity of your MS results.
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Needle Gauge 26 G
Diameter (English) Inner 0.0043 in.
Diameter (English) Outer 0.018 in.
Diameter (Metric) Inner 0.11 mm
Diameter (Metric) Outer 0.45 mm
Includes Ultra Smooth Hydrophilic Coating
Length (English) Needle 2 in.
Product Type GC Needle
Needle Point Style Bevel
For Use With (Equipment) Manual Syringe (10 to 50μL)