Learn More
Trajan™ Diamond LC Needles for 100 to 500μL Manual and Autosampler Syringes
Product Code. 13167906
Click to view available options
Diameter (Metric) Inner:
0.17 mm
0.37 mm
Length (English) Needle:
2 in.
2.24 in.
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Unit Size:
Each
Product Code. 13167906
Brand: Trajan™ 0355221
Removable needle design ensures carryover levels are reduced even lower than the best fixed needle syringes available. Trajan Scientific and Medical™ Diamond LC Needles for 10 to 50μL Manual and Autosampler Syringes are MS ready and designed to improve the sensitivity of your MS results.
Improve the sensitivity of your MS analysis:
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Material Stainless Steel
Needle Gauge 22 G
Diameter (English) Inner 0.0066 in.
Diameter (English) Outer 0.02 in.
Diameter (Metric) Inner 0.17 mm
Diameter (Metric) Outer 0.72 mm
Includes Ultra Smooth Hydrophobic Coating
Length (English) Needle 2 in.
Product Type Diamond Needle
Needle Point Style LC
For Use With (Equipment) Manual and Autosampler Syringes (100 to 500μL)