Learn More
Trajan™ Diamond MS Thermo RSH and CTC RTC Autosampler Syringes
Product Code. 13107986
Click to view available options
Volume (Metric):
10 μL
25 μL
50 μL
100 μL
Model:
100R-RTC/RSH-GT-MS1
10R-RTC/RSH-GT-MS1
25R-RTC/RSH-GT-MS1
50R-RTC/RSH-GT-MS1
Unit Size:
Each
Product Code. 13107986
Brand: Trajan™ 002790
Improves the sensitivity of your MS analysis with reduced non-specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non-specific sample interaction and increased analyte recovery
  • Compatible with ultra smooth removable needles with choice of surface treatment (sold separately)
Specifications
For Use With (Application) Gas/Liquid chromatography
Product Type Autosampler Syringe
Max. Temperature (English) 662°F
Max. Temperature (Metric) 350°C
Model 10R-RTC/RSH-GT-MS1
Quantity Ea.
Volume (Metric) 10 μL
For Use With (Equipment) Thermo RSH and CTC RTC
Connection Type Removable