Learn More
Trajan™ Diamond Thermo RSH 10 to 50μL GC Autosampler Syringes
Product Code. 13117946
Click to view available options
Diameter (Metric) Inner Needle:
0.11 mm
0.24 mm
Includes:
Ultra Smooth Coating
Ultra Smooth Hydrophilic Coating
Ultra Smooth Hydrophobic Coating
Unit Size:
Each
Product Code. 13117946
Brand: Trajan™ 0355610
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Needle Gauge 23 G
Diameter (English) Inner Needle 0.009 in.
Diameter (Metric) Inner Needle 0.24 mm
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Includes Ultra Smooth Hydrophilic Coating
Length (English) Needle 2.24 in.
Product Type GC Autosampler Syringe
Quantity Ea.
Volume (Metric) 10 to 50 μL
Needle Point Style Cone
For Use With (Equipment) Diamond Thermo RSH