Learn More
Trajan™ Diamond Thermo RSH 100μL GC Autosampler Syringes
Product Code. 13127946
Click to view available options
Needle Point Style:
Ultra Smooth Hydrophilic, Cone
Ultra Smooth Hydrophobic, Cone
Ultra Smooth, Cone
Unit Size:
Each
Product Code. 13127946
Brand: Trajan™ 0355611
Improves the sensitivity of your MS analysis with reduced non specific sample interaction and increased analyte recovery.
  • Lowest syringe carryover
  • Reduced non specific sample interaction and increased analyte recovery
  • Ultra smooth removable needles with choice of surface treatment.
Specifications
Needle Gauge 23 G
Diameter (English) Inner Needle 0.009 in.
Diameter (Metric) Inner Needle 0.24 mm
Diameter (English) Outer Needle 0.024 in.
Diameter (Metric) Outer Needle 0.63 mm
For Use With (Application) Gas chromatography
Length (English) Needle 2.24 in.
Product Type Autosampler Syringe
Quantity 1 Pk.
Volume (Metric) 100 μL
Needle Point Style Ultra Smooth Hydrophilic, Cone
For Use With (Equipment) CTC PAL GC