Learn More
Trajan™ eVol™ XCHANGE Syringes
Product Code. 12977586
Click to view available options
Volume (Metric):
100 μL
1 mL
Quantity:
1 Pk.
3/Pk.
Unit Size:
Each
Pack of 3
Product Code. 12977586
Brand: Trajan™ undefined
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available
Specifications
Needle Gauge 25 G
For Use With (Application) Liquid handling procedure
Length (English) 1.9 in.
Length (Metric) 50 mm
Product Type eVol™ Syringe
Quantity 1 Pk.
Volume (Metric) 100 μL
Length (Metric) Needle 50 mm
Needle Point Style Bevel