Learn More
Trajan™ BP5 GC Capillary Column
A general purpose phase used for a wide range of applications
Brand: Trajan™ 054160
Additional Details : Weight : 0.01000kg
Description
- Excellent general purpose column
- Low bleed
- Non-polar
- High temperature
- Ideal for aromatics, pesticides, herbicides, drugs of abuse and hydrocarbons
Specifications
0.53 mm | |
Pi Bonding with the Aromatic Phenyl Groups | |
350/360°C | |
-40°C to +350/360°C | |
30 m | |
Non polar |
G27, G36, G41 | |
5% Phenyl Polysilphenylene Siloxane | |
Phenols Analysis, Ultra Trace Analyses, Pesticides and Herbicides Analysis, Hydrocarbons, Solvents Analysis, Phenols, Amines, GC-MS and Other Specific Detector Applications | |
3 μm | |
Forte™ BPX5 | |
Fused Silica |