Learn More
Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Brand:  Trajan™ 073560
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 16692632
  • € 184.00 / Each
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
  • Low thermal mass
  • Zero Dead Volume
  • Inert
  • No-need to retighten after installation
  • Connect Pre-column or GC x GC
Specifications
Specifications
Connector
0.1-0.25 mm and 0.1-0.25 mm
2 unions, 5 double taper ferrules and tools
GC Capillary Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers