Learn More
Trajan™ GC Capillary Column Connectors: Replacement Ferrules
Click to view available options
For Use With (Equipment):
0.1 to 0.25 mm I.D.
0.1 to 0.25 mm I.D., 0.32 mm I.D.
0.1 to 0.25 mm I.D., 0.53 mm I.D.
0.32 mm I.D.
0.32 mm I.D., 0.53 mm I.D.
0.53 mm I.D.
Unit Size:
Pack of 10
Description
- SilTite μ-Union gives low thermal mass - 9mm in length and <0.5g mass
- Zero dead volume for optimized peak shape
- Chemically deactivated stainless steel ferrules give system inertness
- Robust and stable for both high temperature and pressure cycling
Specifications
Specifications
| Type | Replacement μ-Union Ferrule |
| Material | Stainless Steel |
| For Use With (Equipment) | 0.1 to 0.25 mm I.D., 0.53 mm I.D. |