Learn More
Trajan™ HT8-PCB 8% Phenyl Capillary Columns
Product Code. 10793364
Click to view available options
Length (Metric):
60 m
Unit Size:
Each
Product Code. 10793364
Brand: Trajan™ 054236
Only null left
Add to basket
Add to basket
SGE GC column HT8 is a unique, high temperature phase designed for the analysis of polychlorinated biphenyl (PCB) congeners
  • High temperature, low bleed column
  • Preferred column for polychlorinated biphenyl (PCB) analysis
  • Separates PCBs on ortho ring as well as boiling point
Specifications
Diameter (Metric) Inner 0.25 mm
Stationary Phase SGE™ HT8-PCB
For Use With (Application) GC/MS
Particle Size 0.25 μm
Temperature Range 360°C to 370°C
Film Thickness 0.25 μm
Length (Metric) 60 m