Learn More
Trajan™ Replacement Kit for Plunger-in-Needle Syringes
Product Code. 15248583
Click to view available options
Diameter (Metric) Outer Needle:
0.47 mm
0.63 mm
For Use With (Equipment):
6.5 mm OD Syringe (0.5μL)
8 mm OD Syringe (1μL)
Syringes (1μL)
Unit Size:
Each
Product Code. 15248583
Brand: Trajan™ 034056
SGE NanoVolume syringes are designed to inject down to 50nL with high precision and accuracy
  • High precision and accuracy for small volume injections
  • Replacement plunger/needle kits
  • Kits are matched. Needles and plungers must be replaced as a set
Specifications
Material Stainless Steel, Stainless Steel (Tip)
Needle Gauge 23 G
Diameter (Metric) Outer 0.63 mm
Diameter (Metric) Outer Needle 0.63 mm
Includes Matched plunger and needle
Product Type Replacement Needle and Plunger: NP1B-5BV
Volume (Metric) 1 μL
Needle Point Style Bevel
For Use With (Equipment) 8 mm OD Syringe (1μL)