Learn More
SGE™ Replacement Needles for 0.5 - 1μL Syringes
For use with 0.5 to 1μL syringes
€ 63.70 - € 74.10
Product Code | Brand | For Use With (Equipment) | Needle Gauge | Length (English) Needle | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Needle Gauge | Length (English) Needle | Price | Quantity & Availability | |||
15298573
|
SGE™
033605 |
8 mm O.D. Barrel 0.5μL Microvolume Syringes | 32 G | 4 in. |
€ 72.80
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10147310
|
SGE™
033610 |
8 mm O.D. Barrel 0.5μL Microvolume Syringes | 32 G | 4 in. |
€ 63.70
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10599072
|
SGE™
033630 |
On-column Syringe | 26 G | 2.7 in. |
€ 63.70
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11982595
|
SGE™
034060 |
8 mm O.D. Barrel 1μL Microvolume Syringes | 23 G | 2.7 in. |
€ 74.10
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
10238580
|
SGE™
034720 |
Microvolume Syringes (1μL) | 26 G | 2 in. |
€ 64.90
Each |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15228583
|
SGE™
033772 |
Syringe Needles (0.5μL), Syringes | 23 G | 2 in. |
€ 72.80
Each |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
Description
For removable-needle syringes or additional replacements, contact your Fisher Scientific Customer Service Representative.
Specifications
Stainless Steel | |
Replacement Needle |