Learn More
Trajan™ Replacement Needles with Dome Point
Quality needles to fit SGE syringes
Brand:  Trajan™ 039827
Additional Details : Weight : 0.80000kg
Product Code. 11979427
  • € 63.40 / Pack of 5
Estimated Shipment: 04-06-2024
to see stock.
Description
Description
  • Needle length is the exposed length of needle
  • Choose different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
19 G
0.042 in.
1.07 mm
2.7 in.
5/Pk.
Bevel
0.025 in.
0.65 mm
Dome point
Luer Lock Needle
70 mm