Learn More
Trajan™ Replacement Plungers
Click to view available options
For Use With (Equipment):
10 μL Gas-tight Autosampler Fixed Needle Syringe
10 μL Removable Guided Plunger Syringe
10 μL Removable Needle Gas-tight Syringe for CTC/Carlo Erba/Fisons Autosampler
100 μL Fixed Needle Gas-tight Syringe for Carlo Erba, CTC, Fisons Autosamplers
Fixed Needle Gas Tight Syringe (10μL)
P250-GT Code Syringe (250μL)
Removable Fitting Gas Tight Syringe (10 mL)
Removable Needle Gas Tight Syringe (10μL)
Rheodyne and Valco Valves
Syringe 009472, 009462
Syringe 100μL
Syringe Models SG-003987, SG-003988, 25 μL
Syringe Models SG-003988, SG-003985, 25 μL
Syringe Needles (10 μL)
Syringe Needles (10 μL), Fixed Needle Gas Tight Syringe
Syringe Needles (100μL), Gas Tight Syringe
Syringe Needles (100μL), Removable Needle Gas Tight Syringe
Syringe Needles (25μL), CTC Gas Tight Syringe
Syringe Needles (25μL), Fixed Needle Gas Tight Syringe
Syringe Needles (25μL), Fixed or Removable Needle Gas Tight Syringe
Syringe Needles (250μL), CTC Gas Tight Syringe
Syringe Needles (250μL), Gas Tight Syringe
Syringe Needles (5 mL), Gas Tight Syringe
Syringe Needles (5μL), Fixed Needle Gas Tight Syringe
Syringe Needles (50μL), Fixed Needle Gas Tight Syringe
Syringe Needles (500μL), CTC Gas Tight Syringe
Syringe Needles (500μL), Fixed or Removable Needle Gas Tight Syringe
Product Type:
50 mL Replacement Plunger only
Plunger
Replacement Plunger (old style), P25F-BT-GT
Replacement Plunger, 10A-VP
Replacement Plunger, 10R-C/T-GT
Replacement Plunger, P10-GP-GT
Replacement Plunger, P100-GT
Replacement Plunger, P100F-C/T-GT
Replacement Plunger, P100R-C/T-GT
Replacement Plunger, P10F-BT-GT
Replacement Plunger, P10F-C/T-GT
Replacement Plunger, P10MD-GT
Replacement Plunger, P10R-GT
Replacement Plunger, P25-GT
Replacement Plunger, P250F-CTC-GT
Replacement Plunger, P250R-C/T-GT
Replacement Plunger, P25F-C/T-GT
Replacement Plunger, P25F-CTC-GT
Replacement Plunger, P25MD-GT
Replacement Plunger, P25R-C/T-GT
Replacement Plunger, P500-GT
Replacement Plunger, P500F-CTC-GT
Replacement Plunger, P50F-C/T-GT
Replacement Plunger, P5F-PE-GT
Replacement Plunger, P5MD-GT
Unit Size:
Each
Pack of 2
Description
- For use with SGE gas tight syringes.
- Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
Specifications
| Material | PTFE (Tip) |
| For Use With (Application) | IN VITRO use only. For lab analyzers only. Not for human use. |
| Product Type | Replacement Plunger, P10F-BT-GT |
| For Use With (Equipment) | Fixed Needle Gas Tight Syringe (10μL) |