Learn More
SGE™ SilFlow™ GC Replacement Parts
Product Code. 15299163
Click to view available options
Description:
Ferrule, 0.36 mm
Ferrule, 0.5 mm
Ferrule, 0.7 mm
Ferrule, 1.1 mm
Finger Tite Tool
Nut
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 15299163
Brand: SGE™ 123705
Only null left
Add to basket
Add to basket
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
  • Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
Specifications
Type Ferrule
Description Ferrule, 1.1 mm
For Use With (Equipment) SGE Silflow Microchannel Device
Diameter (Metric) Inside 1.1 mm