Learn More
SGE™ SilFlow™ GC Replacement Parts
Product Code. 15209173
Change view
Click to view available options
Description:
Ferrule, 0.36 mm
Ferrule, 0.5 mm
Ferrule, 0.7 mm
Ferrule, 1.1 mm
Finger Tite Tool
Nut
Unit Size:
Each
Pack of 10
Pack of 5
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
15209173 Ferrule, 0.36 mm Pack of 10
15299163 Ferrule, 1.1 mm Pack of 5
15289163 Nut Pack of 10
15239173 Ferrule, 0.7 mm Pack of 10
15219173 Ferrule, 0.5 mm Pack of 10
15279173 Finger Tite Tool Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15209173 Supplier SGE™ Supplier No. 123706
Only null left
Add to basket
Add to basket
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
  • Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
Specifications
Type Ferrule
Description Ferrule, 0.36 mm
For Use With (Equipment) SGE Silflow Microchannel Device
Diameter (Metric) Inside 0.36 mm