Learn More
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
Brand:  SGE™ 073224
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12790740
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
  • Non-Swagelok™ nuts with two notches on outer surface
  • Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
  • Ferrules are easily removed from nut while still attached to capillary column
  • Reusable
Specifications
Specifications
SilTite Metal Ferrule
Split/Splitless Injector
Nut
Used for GC/MS Interface Connections
Metal
5/Pk.