missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SgK071 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00169436-B01P
This item is not returnable.
View return policy
Description
SgK071 Polyclonal antibody specifically detects SgK071 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SgK071 | |
| Polyclonal | |
| Western Blot 1:500, Immunocytochemistry/ Immunofluorescence | |
| AAH36504.3 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| chromosome 9 open reading frame 96, MGC43306, protein kinase-like protein SgK071, RP11-244N20.8, SgK071, Sk521, Sugen kinase 071 | |
| C9orf96 (AAH36504.3, 1 a.a. - 680 a.a.) full-length human protein. MLGPGSNRRRPTQGERGPGSPGEPMEKYQVLYQLNPGALGVNLVVEEMETKVKHVIKQVECMDDHYASQALEELMPLLKLRHAHISVYQELFITWNGEISSLYLCLVMEFNELSFQEVIEDKRKAKKIIDSEWMQNVLGQVLDALEYLHHLDIIHRNLKPSNIILISSDHCKLQDLSSNVLMTDKAKWNIRAEEDPFRKSWMAPEALNFSFSQKSDIWSLGCIILDMTSCSFMDGTEAMHLRKSLRQSPGSLKAVLKTMEEKQIPDVETFRNLLPLMLQIDPSDRITIKDVVHITFLRGSFKSSCVSLTLHRQMVPASITDMLLEGNVASILEVMQKFSGWPEVQLRAMKRLLKMPADQLGLPWPPELVEVVVTTMELHDRVLDVQLCACSLLLHLLGQALVHHPEAKAPCNQAITSTLLSALQSHPEEEPLLVMVYSLLAITTTQESESLSEELQNAGLLEHILEHLNSSLESRDVCASGLGLLWALLLDGIIVNKAPLEKVPDLISQVLATYPADGEMAEASCGVFWLLSLLGCIKEQQFEQVVALLLQSIRLCQDRALLVNNAYRGLASLVKVSELAAFKVVVQEEGGSGLSLIKETYQLHRDDPEVVENVGMLLVHLASYEEILPELVSSSMKALLQEIKERFTSSLVSDSSAFSKPGLPPGGSPQLGCTTSGGLE | |
| 0.05 mg | |
| Protein Kinase | |
| 169436 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction