missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98778AF532
This item is not returnable.
View return policy
Description
SGK1 Polyclonal specifically detects SGK1 in Human samples. It is validated for Western Blot.
Specifications
| SGK1 | |
| Polyclonal | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human SGK1. | |
| 0.1 mL | |
| Apoptosis, DNA Repair | |
| 6446 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction