Learn More
SGK196 Mouse anti-Human, Clone: OTI1B1, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF804623
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The exact function of FLJ23356 remains unknown.Specifications
| SGK196 | |
| Monoclonal | |
| Unconjugated | |
| POMK | |
| MDDGA12, MDDGC12, SGK196 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84197 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1B1 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H5K3 | |
| POMK | |
| Human recombit protein fragment corresponding to amino acids 141-350 of human SGK196 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.