missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGLT1/SLC5A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 369.00 - € 593.00
Specifications
| Antigen | SGLT1/SLC5A1 |
|---|---|
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18415112
|
Novus Biologicals
NBP2-33629-25ul |
25 μL |
€ 369.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18746393
|
Novus Biologicals
NBP2-33629 |
0.1 mL |
€ 593.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGLT1/SLC5A1 Polyclonal specifically detects SGLT1/SLC5A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| SGLT1/SLC5A1 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| P13866 | |
| 6523 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: DLDAEEENIQEGPKETIEIETQVPEKKKGIFRRAYDLFCGLEQHGAPKMTEEEEKAMKMKMTDTSEKPLWR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Polyclonal | |
| Rabbit | |
| Membrane Trafficking and Chaperones | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| D22S675, High affinity sodium-glucose cotransporter, Na+/glucose cotransporter 1, NAGTsodium/glucose cotransporter 1, SGLT1Na(+)/glucose cotransporter 1, solute carrier family 5 (sodium/glucose cotransporter), member 1, Solute carrier family 5 member 1 | |
| SLC5A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title