missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGMS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598586
This item is not returnable.
View return policy
Description
SGMS1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
A bidirectional lipid cholinephosphotransferases capable of converting phosphatidylcholine (PC) and ceramide to sphingomyelin (SM) and diacylglycerol (DAG) and the inverse reaction. Direction is dependent on the relative concentrations of DAG and ceramide as phosphocholine acceptors. Directly and specifically recognizes the choline head group on the substrate. Also requires two fatty chains on the choline-P donor molecule in order to be recognized efficiently as a substrate. May not function strictly as a SM synthase. Inhibited by bacterial PC-phospholipase C inhibitor D609. Localized to Golgi apparatus; Golgi membrane; multi-pass membrane protein. Possibly present on Brain, heart, kidney, liver, muscle and stomach.
Specifications
| SGMS1 | |
| Polyclonal | |
| Unconjugated | |
| SGMS1 | |
| 9530058O11Rik; AI841905; C80702; hmob33; Medulla oblongata-derived protein; MGC17342; MOB; MOB1; mob-1; Phosphatidylcholine:ceramide cholinephosphotransferase 1; protein Mob; SGMS1; SMS1; Sor1; sphingomyelin synthase 1; TMEM23; Transmembrane protein 23 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 259230 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4.24 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q86VZ5 | |
| SGMS1 | |
| Recombinant Human Phosphatidylcholine:ceramide cholinephosphotransferase 1 protein (48-137AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction