missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGPL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121614
This item is not returnable.
View return policy
Description
Sphingosine-1-phosphate lyase cleaves phosphorylated sphingoid bases, such as sphingosine-1 phosphate, into fatty aldehydes and phosphoethanolamine. Elevates stress-induced ceramide production and apoptosis. The catalytic activity of sphingosine 1- phosphate lyase converts sphingosine 1-phosphate to phosphoethanolamine and palmitaldehyde using Pyridoxal phosphate as a co-factor. Sphingosine 1-phosphate lyase is a type III membrane protein found in the endoplasmic reticulum. Sphingosine 1-phosphate lyase is found primarily in the liver and kidney. Sphingosine 1-phosphate lyase belongs to the group II decarboxylase family.
Specifications
| SGPL1 | |
| Polyclonal | |
| Unconjugated | |
| SGPL1 | |
| AI428538; D10Xrf456; FLJ13811; hSPL; KIAA1252; mSPL; S1PL; Sgpl1; Spgl1; sphingosine phosphate lyase 1; sphingosine-1-phosphate aldolase; Sphingosine-1-phosphate lyase 1; sphingosine-1-phosphate lyase 1-like protein; spl; SPL 1; SP-lyase 1; zgc:162153 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 8879 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O95470 | |
| SGPL1 | |
| Gln239-Arg493 (Accession O95470), with N-terminal His Tag. | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction