missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SGSM1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15759990 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15759990 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15759990 Supplier Invitrogen™ Supplier No. PA552038

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SPDSGHPSSH NFSSGLSEHS EPSLSTEDSV LDAQRNTPTV LRPRDGSVDD RQSSEATTSQ DEAPREELAV QDSLESDLLA NESMDEFMSI TGSLDMALPE KDDVVMEGWR SSETEKHGQA DSEDNLSEEP EMESLFPALA SLAVTT Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.

Small G proteins such as RAP and RAB proteins are the key molecules in intracellular signal transduction and vesicle transportation. A novel protein family small G protein signaling modulator (SGSM) consisting of three members SGSM1-3 bind to RAP and RAB family proteins. All three SGSM proteins possess both a RUN domain and a TBC domain. SGSM1 (RUTBC2) is a 1, 148 amino acid protein that localizes to the Golgi apparatus and is mainly expressed in the CNS. SGSM1 interacts with RAP and RAB subfamily members of the small G proteins, and function as modulators of RAP and RAB-mediated neuronal signal transduction and vesicular transportation pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SGSM1
Gene Accession No. Q2NKQ1
Gene Alias 2410098H20Rik; AA408781; D5Bwg1524e; E130107M15; KIAA1941; NuIP; nurr1-interacting protein; RUN and TBC1 domain containing 2; RUN and TBC1 domain-containing protein 2; RUTBC2; SGSM1; Small G protein signaling modulator 1; small G protein signaling modulator 1 protein
Gene Symbols SGSM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SGSM1. Recombinant protein control fragment (Product #RP-91520).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 129049
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.