missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGSM3 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18448DL594
This item is not returnable.
View return policy
Description
SGSM3 Polyclonal antibody specifically detects SGSM3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| SGSM3 | |
| Polyclonal | |
| Western Blot | |
| DJ1042K10.2, DKFZp761D051, MAP, merlin binding protein, Merlin-associated protein, rabGAPLP, Rab-GTPase-activating protein-like protein, RUN and SH3 containing 3, RUN and TBC1 domain containing 3, RUN and TBC1 domain-containing protein 3, RUSC3, RUTBC3, small G protein signaling modulator 3, small G protein signaling modulator 3 protein | |
| Recognizes region between residues Between Between 699 and 749.. | |
| 0.1 mL | |
| Signal Transduction | |
| 27352 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction