missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2B1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12226-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the SH2-domain containing mediators family. The encoded protein mediates activation of various kinases and may function in cytokine and growth factor receptor signaling and cellular transformation. Alternatively spliced transcript variants have been described.Specifications
| SH2B1 | |
| Polyclonal | |
| Unconjugated | |
| SH2B1 | |
| KIAA1299, PSM, SH2B, SH2B adapter protein 1, SH2B adaptor protein 1, SH2B1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 25970 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9NRF2 | |
| SH2B1 | |
| SH2B1 Fusion Protein Ag2863 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction