missing translation for 'onlineSavingsMsg'
Learn More

SH2B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-22515-0.025mg

 View more versions of this product

Product Code. 18763014

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

SH2B2 Polyclonal specifically detects SH2B2 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SH2B2
Polyclonal
Western Blot 1:500
adaptor protein with pleckstrin homology and src homology 2 domains, APSAdapter protein with pleckstrin homology and Src homology 2 domains, SH2 and PH domain-containing adapter protein APS, SH2B adapter protein 2, SH2B adaptor protein 2
Rabbit
Affinity Purified
RUO
10603
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
PBS with 1 mg/mL BSA with 0.05% Sodium Azide
SH2B2
Synthetic peptide corresponding to residues E(605) A T L G R A R A V E N Q Y S(619) from mouse APS.
0.025 mg
Primary
Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only