Learn More
SH2B3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22198-1-AP-20UL
 View more versions of this product
Product Code. 16811735
  • € 145.00 / 20µL
Description
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
Specifications
Specifications
SH2B3
Polyclonal
Unconjugated
SH2B3
LNK, Lymphocyte adapter protein, SH2B adapter protein 3, SH2B adaptor protein 3, SH2B3
Rabbit
Antigen Affinity Chromatography
RUO
10019
-20°C
Liquid
Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9UQQ2
SH2B3
SH2B3 Fusion Protein Ag17381