missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ SH2D1A Antibody (08) [DyLight 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26884UV
This item is not returnable.
View return policy
Description
SH2D1A Monoclonal antibody specifically detects SH2D1A in Human samples. It is validated for Sandwich ELISA Capture
Specifications
| SH2D1A | |
| Monoclonal | |
| DyLight 350 | |
| DSHPLYP, Duncan disease SH2-protein, EBVS, FLJ92177, IMD5, MTCP1, SAP/SH2D1A, SAPlymphoproliferative syndrome, SH2 domain containing 1A, SH2 domain-containing protein 1A, signaling lymphocyte activation molecule-associated protein, Signaling lymphocytic activation molecule-associated protein, SLAM associated protein/SH2 domain protein 1A, SLAM-associated protein, T cell signal transduction molecule SAP, T-cell signal transduction molecule SAP, XLPDFLJ18687, XLPSH2 domain protein 1A | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human SH2D1A (RefSeq#: NP_002342.1; Pro2-Lys97). | |
| 0.1 mL | |
| Immune System Diseases, Immunology | |
| 4068 | |
| Store at 4°C in the dark. | |
| IgG |
| Sandwich ELISA Capture | |
| 08 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction