missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH2D1B Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA563020

Product Code. 15750802

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LPYYHGRLTK QDCETLLLKE GVDGNFLL Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 75%.

SH2D1B plays a role in controlling signal transduction through at least four receptors, CD84, CD150, CD229 and CD244, expressed on the surface of professional antigen-presenting cells.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SH2D1B
Polyclonal
Unconjugated
Sh2d1b
Eat2; EAT-2; Eat2a; EAT-2A; EWS/FLI1 activated transcript 2; EWS/FLI1-activated transcript 2; RGD1563935; SH2 domain containing 1B; SH2 domain containing 1B1; SH2 domain protein 1B1; SH2 domain-containing molecule EAT2; SH2 domain-containing protein 1B; Sh2d1b; Sh2d1b1
Rabbit
Antigen affinity chromatography
RUO
117157
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14796
Sh2d1b
Recombinant protein corresponding to Human SH2D1B. Recombinant protein control fragment (Product #RP-101476).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.