missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH2D2A Antibody (OTI4F3), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74153PEATT594
This item is not returnable.
View return policy
Description
SH2D2A Monoclonal specifically detects SH2D2A in Human samples. It is validated for Flow Cytometry.
Specifications
| SH2D2A | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| SH2D2A | |
| Full length recombinant protein of human SH2D2A (NP_003966) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| OTI4F3 | |
| Flow Cytometry | |
| F2771, SCAP, SH2 domain containing 2A, SH2 domain protein 2A, SH2 domain-containing adapter protein, T cell specific adapter protein TSAd, T cell-specific adapter protein, T lymphocyte specific adaptor protein, TSAd, TSADSH2 domain-containing protein 2A, VEGF receptor-associated protein, VRAP | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis | |
| 9047.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction