missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH2D3C Polyclonal Antibody
GREENER_CHOICE

Product Code. 16360825
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16360825 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16360825 Supplier Invitrogen™ Supplier No. PA583872

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SGAIIYCPVN RTFPLRYLEA SYGLGQGSSK PASPVSPSGP KGSHMKRRSV TMTDGLTADK VTRSDGCPTS TSLPRPRDSI RSCALSMDQI PDLH Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 91%.

Eph receptor-binding protein which may be a positive regulator of TCR signaling. Binding to BCAR1 is required to induce membrane ruffling and promote EGF-dependent cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SH2D3C
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SH2D3C
Gene Accession No. Q8N5H7
Gene Alias Cas/HEF1-associated signal transducer; CHAT; Novel SH2-containing protein 3; NSP3; PRO34088; SH2 domain containing 3C; SH2 domain-containing Eph receptor-binding protein 1; SH2 domain-containing protein 3C; Sh2d3c; Shep1; UNQ272/PRO309/PRO34088
Gene Symbols SH2D3C
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SH2D3C. Recombinant protein control fragment (Product #RP-92892).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10044
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.