missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH2D4A Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA519055
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1,000.
SH2D4A inhibits estrogen-induced cell proliferation by competing with PLCG for binding to ESR1, blocking the effect of estrogen on PLCG and repressing estrogen-induced proliferation. SH2D4A may play a role in T-cell development and function.
Specifications
| SH2D4A | |
| Polyclonal | |
| Unconjugated | |
| SH2D4A | |
| 2210402M20Rik; FLJ20967; PPP1R38; protein phosphatase 1 regulatory subunit 38; protein phosphatase 1, regulatory subunit 38; protein SH(2)A; SH2 domain containing 4A; SH2 domain-containing 4A; SH2 domain-containing protein 4A; SH2A; SH2D4A | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 63898 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q9H788 | |
| SH2D4A | |
| Synthetic peptide sequence (HQKKESMKKKQDE) corresponding to the internal amino acids of SH2D4A (aa 197-210). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction