missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BGRL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104140
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total SH3BGRL1.
SH3BGRL belongs to the SH3BGR family. Mutations in predicted EVH1-binding domain of SH3BGRL had a modest effect on suppression of v-Rel transformation.
Specifications
| SH3BGRL | |
| Polyclonal | |
| Unconjugated | |
| SH3BGRL | |
| 1190008F14Rik; epididymis secretory protein Li 115; HEL-S-115; RGD1560520; SH3 domain binding glutamate rich protein like; SH3 domain binding glutamate-rich protein like; SH3 domain binding glutamic acid-rich protein like; SH3 domain-binding glutamic acid-rich-like protein; SH3BGR; Sh3bgrl; SH3-binding domain glutamic acid-rich protein like; similar to SH3 domain binding glutamic acid-rich protein like; zgc:64130; zSH3BGRL | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 302363, 56726, 6451 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O75368, Q9JJU8 | |
| SH3BGRL | |
| A synthesized peptide derived from human SH3BGRL(Accession O75368), corresponding to amino acid residues N56-A106. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction