Learn More
Novus Biologicals™ SH3BP2 Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30802-50ug
 View more versions of this product
Product Code. 30067058
  • € 213.00 / 50µg
Description
SH3BP2 Polyclonal antibody specifically detects SH3BP2 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SH3BP2
Polyclonal
Phosphate buffered saline
Sheep
Ammonium sulfate precipitation
RUO
Primary
Mouse
Purified
Western Blot
Unconjugated
Abl-SH3 binding protein 2, Cherubism, CRBM, CRPM, FLJ42079, FLJ54978,3BP-2,3BP2, RES4-23, SH3 domain-binding protein 2, SH3-domain binding protein 2, TNFAIP3 interacting protein 2
Antibody developed using SH2 domain of the SH3BP2 protein fused to GST.
50 μg
Cell Biology, Phospho Specific, Signal Transduction
6452
Store at -20°C. Avoid freeze/thaw cycles.