missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3BP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA529031
This item is not returnable.
View return policy
Description
Recommended positive controls: A431, HeLa, HepG2, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene has an N-terminal pleckstrin homology (PH) domain, an SH3-binding proline-rich region, and a C-terminal SH2 domain. The protein binds to the SH3 domains of several proteins including the ABL1 and SYK protein tyrosine kinases, and functions as a cytoplasmic adaptor protein to positively regulate transcriptional activity in T, natural killer (NK), and basophilic cells. Mutations in this gene result in cherubism. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| SH3BP2 | |
| Polyclonal | |
| Unconjugated | |
| Sh3bp2 | |
| 3BP2; 3BP-2; Abl-SH3 binding protein 2; CRBM; CRPM; FLJ42079; LOW QUALITY PROTEIN: SH3 domain-binding protein 2; RES4-23; SH3 domain binding protein 2; SH3 domain-binding protein 2; SH3BP2; SH3-domain binding protein 2; TNFAIP3 interacting protein 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6452 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| P78314 | |
| Sh3bp2 | |
| Recombinant protein encompassing a sequence within the center region of human 3BP2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction