missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SH3BP4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA557993

Product Code. 15775881

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LKVCMFSNMT NYEVKASEQA KVVRGFQLKL GKVSRLIFPI TSQNPNELSD FTLRVQVKDD QEAILTQFCV QTPQPPPKSA IKPSGQRRFL KKNEVGKIIL Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

SH3BP4 was initially isolated as a novel cDNA using differential display analysis using cultured human corneal fibroblasts. The protein contains three Asn-Pro-Phe (NPF) motifs, an SH3 domain, a PXXP motif, a bipartite nuclear targeting signal, and a tyrosine phosphorylation site. SH3BP4 has been shown to interact with specific endocytic proteins such as clathrin, dynamin, and the transferrin receptor (TfR) and localizes to TfR-containing coated pits and vesicles. It is thought to be involved in cargo-specific control of clathrin-mediated endocytosis, specifically controlling the internalization of TfR. At least two isoforms of SH3BP4 are known to exist.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SH3BP4
Polyclonal
Unconjugated
SH3BP4
AI594717; AW227605; BOG25; EHB10; EH-binding protein 10; SH3 domain binding protein 4; SH3 domain-binding protein 4; SH3 domain-binding protein 4; SH3-domain binding protein 4; Sh3bp4; SH3-domain binding protein 4; si:dkey-57b6.3; Transferrin receptor-trafficking protein; TTP
Rabbit
Antigen Affinity Chromatography
RUO
23677
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9P0V3
SH3BP4
Recombinant protein corresponding to Human SH3BP4. Recombinant protein control fragment (Product #RP-102773).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.