missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3BP4 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | SH3BP4 |
|---|---|
| Concentration | 600 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30234225
|
Proteintech
29623-1-AP-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236630
|
Proteintech
29623-1-AP-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SH3BP4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SH3BP4. This antibody recognizes human, mouse , rat antigen. The SH3BP4 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| SH3BP4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| BOG25; TTP | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 600 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 23677 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts