missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL2, Mouse anti-Human, Clone: 5A6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00006456-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: SRAKLSMINTMSKIRGQEKGPGYPQAEALLAEAMLKFGRELGDDCNFGPALGEVGEAMRELSpecifications
| SH3GL2 | |
| Monoclonal | |
| Unconjugated | |
| SH3-domain GRB2-like 2 | |
| CNSA2/EEN-B1/FLJ20276/FLJ25015/SH3D2A/SH3P4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 5A6 | |
| In 1x PBS, pH 7.4 | |
| NM_003026 | |
| SH3GL2 | |
| SH3GL2 (NP_003017, 64 a.a. ∼ 124 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 6456 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction