missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3GL2 Polyclonal Antibody

SH3GL2 Polyclonal Antibody
Brand: Invitrogen PA5101888
This item is not returnable.
View return policy
Description
SH3GL2 Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| SH3GL2 | |
| Polyclonal | |
| Unconjugated | |
| Sh3gl2 | |
| 9530001L19Rik; AI120490; AW555077; B930049H17Rik; bA335L15.1; bA335L15.1 (SH3-domain GRB2-like 2); CNSA2; Een1; EEN-B1; endophilin; endophilin A1; Endophilin A1 BAR domain; endophilin I; endophilin-1; Endophilin-A1; FLJ20276; FLJ25015; SH3 domain containing GRB2 like 2; SH3 domain containing GRB2 like 2, endophilin A1; SH3 domain protein 2A; SH3 domain-containing GRB2-like protein 2; Sh3d2a; SH3-domain GRB2-like 2; SH3GL2; Sh3p4; SH3PA | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 116743, 20404, 6456 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35179, Q62420, Q99962 | |
| Sh3gl2 | |
| A synthesized peptide derived from human SH3GL2(Accession Q99962), corresponding to amino acid residues M51-G101. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction