missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SH3PX1 Antibody (OTI1B5), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74158H
This item is not returnable.
View return policy
Description
SH3PX1 Monoclonal specifically detects SH3PX1 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SH3PX1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| SNX9 | |
| Full length recombinant protein of human SH3PX1 (NP_057308) produced in HEK293T cells. | |
| 0.1 mL | |
| 51429.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1B5 | |
| Western Blot, Immunohistochemistry | |
| MST155, MSTP155, Protein SDP1, SDP1, SH3 and PX domain-containing protein 1, SH3PX1WISP, SH3PXD3ASH3 and PX domain-containing protein SH3PX1, sorting nexin 9, sorting nexin-9, WiskSH3 and PX domain-containing protein 3A | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Primate, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction