missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SH3PXD2B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144870
This item is not returnable.
View return policy
Description
Immunogen sequence: ASRPNFLAPL PHEVTQLRLG EAAALENNTG SEATGPSRPL PDAPHGVMDS GLPWSKDWKG SKDVLRKASS DMSASAGYEE ISDPDMEEKP SLPPRKESII KS Antigen sequence identity: Mouse 82%; Rat 83%.
Adapter protein involved in invadopodia and podosome formation and extracellular matrix degradation. Binds matrix metalloproteinases (ADAMs), NADPH oxidases (NOXs) and phosphoinositides. Acts as an organizer protein that allows NOX1- or NOX3-dependent reactive oxygen species (ROS) generation and ROS localization. Plays a role in mitotic clonal expansion during the immediate early stage of adipocyte differentiation.
Specifications
| SH3PXD2B | |
| Polyclonal | |
| Unconjugated | |
| SH3PXD2B | |
| adapter protein HOFI; adaptor protein HOFI; factor for adipocyte differentiation 49; FAD49; FLJ20831; FTHS; HOFI; KIAA1295; SH3 and PX domain-containing protein 2B; SH3 and PX domains 2B; SH3PXD2B; TKS4; TSK4; tyrosine kinase substrate with four SH3 domains | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 285590 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1X283 | |
| SH3PXD2B | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 431-532. Recombinant protein control fragment (Product #RP-110002). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction